missing translation for 'onlineSavingsMsg'
Learn More

Azelnidipine 98.0+%, TCI America™

Catalog No. A24335G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24335G 5 g
A24331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24335G Supplier TCI America Supplier No. A24335G
Only null left

Chemical Identifiers

CAS 123524-52-7
Molecular Formula C33H34N4O6
Molecular Weight (g/mol) 582.66
MDL Number MFCD00865803
InChI Key ZKFQEACEUNWPMT-UHFFFAOYNA-N
Synonym 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl Ester, 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate
PubChem CID 65948
IUPAC Name 3-[1-(diphenylmethyl)azetidin-3-yl] 5-propan-2-yl 2-amino-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC(C)OC(=O)C1=C(C)NC(N)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 582.66
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Azelnidipine
RTECSNumber US7968332
TSCA No