missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azelnidipine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 582.66 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Azelnidipine |
Safety and Handling
| RTECSNumber | US7968332 |
| TSCA | No |