missing translation for 'onlineSavingsMsg'
Learn More

Azelastine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015555
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015555 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015555 Supplier Merck Emd Millipore Supplier No. PHR16361G

missing translation for 'validMainframeMsg'

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Azelastine Hydrochloride is a long-acting antiallergic and antiasthmatic drug. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 79307-93-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C22H24 ClN3 O · HCl
MDL Number MFCD00242783
Synonym Azelastine hydrochloride; 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)phthalazin-1-one hydrochloride; Astelin; Optivar
Type Certified Reference Material
Quantity 1 g
Formula Weight 418.36
Grade Certified Reference Material