missing translation for 'onlineSavingsMsg'
Learn More

Azelaic Dihydrazide 95.0+%, TCI America™

Catalog No. A074625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A074625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A074625G Supplier TCI America Supplier No. A074625G
Only null left

Chemical Identifiers

CAS 4080-95-9
Molecular Formula C9H20N4O2
Molecular Weight (g/mol) 216.285
MDL Number MFCD00051529
InChI Key ZWLFGLCGZUVIEA-UHFFFAOYSA-N
PubChem CID 77699
IUPAC Name nonanedihydrazide
SMILES C(CCCC(=O)NN)CCCC(=O)NN

Specifications

Melting Point 183°C
Color White
Quantity 25 g
Formula Weight 216.29
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Azelaic Dihydrazide