accessibility menu, dialog, popup

UserName

H2NNHOONHNH2

CAS RN 4080-95-9

IUPAC Name: nonanedihydrazide
Synonyms: nonanedihydrazide
Molecular Weight (g/mol): 216.29
Molecular Formula: C9H20N4O2
InChi Key: ZWLFGLCGZUVIEA-UHFFFAOYSA-N
SMILES: NNC(=O)CCCCCCCC(=O)NN
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results