missing translation for 'onlineSavingsMsg'
Learn More

Azelaic Acid, Analytical Standard, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012825
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012825 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012825 Supplier Merck Emd Millipore Supplier No. 95054100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 123-99-9
Vapor Pressure <1 mmHg (20°C)
Vapor Density 6.5 (vs. air)
Physical Form Neat
Linear Formula HO2C(CH2)7 CO2H
MDL Number MFCD00004432
Melting Point 106°C to 112°C; 109°C to 111°C
Boiling Point 286°C (100 mmHg)
Synonym Nonanedioic acid
Quantity 100 mg
Formula Weight 188.22
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less