missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azaperone, Thermo Scientific™
Chemical Identifiers
| CAS | 1649-18-9 |
|---|---|
| Molecular Formula | C19H22FN3O |
| Molecular Weight (g/mol) | 327.40 |
| MDL Number | MFCD00866692 |
| InChI Key | XTKDAFGWCDAMPY-UHFFFAOYSA-N |
| Synonym | azaperone, stresnil, fluoperidol, suicalm, azaperon, azeperone, eucalmyl, sedaperone vet, azaperona, azaperonum |
| PubChem CID | 15443 |
| IUPAC Name | 1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one |
| SMILES | FC1=CC=C(C=C1)C(=O)CCCN1CCN(CC1)C1=CC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC460830010
|
Thermo Scientific Chemicals
460830010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC460830050
|
Thermo Scientific Chemicals
460830050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC460830250
|
Thermo Scientific Chemicals
460830250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1649-18-9 | |
| 327.40 | |
| XTKDAFGWCDAMPY-UHFFFAOYSA-N | |
| 15443 | |
| FC1=CC=C(C=C1)C(=O)CCCN1CCN(CC1)C1=CC=CC=N1 |
| C19H22FN3O | |
| MFCD00866692 | |
| azaperone, stresnil, fluoperidol, suicalm, azaperon, azeperone, eucalmyl, sedaperone vet, azaperona, azaperonum | |
| 1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one |
Specifications
| 1649-18-9 | |
| Authentic | |
| MFCD00866692 | |
| azaperone, stresnil, fluoperidol, suicalm, azaperon, azeperone, eucalmyl, sedaperone vet, azaperona, azaperonum | |
| XTKDAFGWCDAMPY-UHFFFAOYSA-N | |
| 1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one | |
| 15443 | |
| Azaperone |
| 0.5% max. (60°C, 4 h) (vacuum) | |
| C19H22FN3O | |
| 1g | |
| Solubility in water: insoluble. Other solubilities: soluble in methanol, chloroform | |
| FC1=CC=C(C=C1)C(=O)CCCN1CCN(CC1)C1=CC=CC=N1 | |
| 327.40 | |
| 327.4 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 216-715-8
RUO â Research Use Only