missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne acid, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals J64549MB

Quantity 25mg

 View more versions of this product

Catalog No. AAJ64549MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Azadibenzocyclooctyne acid is the simplest amine reactive reagent for introducing a dibenzylcyclooctyne to available amine functionality.Azadibenzocyclooctyne acid is widely useful in strain-promoted copper-free azide-alkyne click chemistry reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store in cool place. Incompatible with amines.
Specifications

Chemical Identifiers

1425485-72-8
333.39
NIRLBCOFKPVQLM-UHFFFAOYSA-N
77078156
C21H19NO3
MFCD22380737
dbco-c6-acid, azadibenzocyclooctyne acid, dibenzocyclooctyne-acid, storage temp.:-20c, 6-11,12-didehydro-5,6-dihydrodibenzo b,f azocine-5-yl-6-oxohexanoic acid

Specifications

Azadibenzocyclooctyne acid
C21H19NO3
MFCD22380737
Soluble in dimethyl sulfoxide and dimethyl formamide.
333.39
333.38
1425485-72-8
25mg
dbco-c6-acid, azadibenzocyclooctyne acid, dibenzocyclooctyne-acid, storage temp.:-20c, 6-11,12-didehydro-5,6-dihydrodibenzo b,f azocine-5-yl-6-oxohexanoic acid
NIRLBCOFKPVQLM-UHFFFAOYSA-N
77078156
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions