missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Axypet™ Single-Channel Pipettor

Axygen™ Axypet™ pipettors combine high levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.

Supplier:  Axygen™ CAP200

 View more versions of this product

Catalog No. 14223570


Only null left

Description

Description

  • Thumb-friendly soft spring pipetting system reduces fatigue
  • 3-year warranty
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
Specifications

Chemical Identifiers

1
Axypet
3 years

Specifications

±1.2/ ±0.6%
1
Single-Channel Pipettor
±0.6/ ±0.2%
Tip Type: 200 μL
Axypet
20 to 200 μL
3 years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions