Learn More
Axygen™ Axypet™ Single-Channel Pipettor
Catalog No. 14223569
Change view
Click to view available options
Volume (Metric):
2 to 20 μL
Catalog No. Volume (Metric)
14223569 2 to 20 μL
1 options
Catalog No. 14223569 Supplier Axygen™ Supplier No. AP20
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Axygen™ Axypet™ pipettors combine high levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • 3-year warranty
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
Specifications
Accuracy ±3.0/ ±0.8%
Description Tip Type: 200 μL
Number of Channels 1
Product Line Axypet
Type Single-Channel Pipettor
Volume (Metric) 2 to 20 μL
Precision ±1.5/ ±0.3%
Warranty 3 years