missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Axypet™ Single-Channel Pipettor
GREENER_CHOICE

Axygen™ Axypet™ pipettors combine high levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.

Supplier:  Axygen™ AP1000

 View more versions of this product

Catalog No. 14223562


Only null left

Description

Description

  • Thumb-friendly soft spring pipetting system reduces fatigue
  • 3-year warranty
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
Specifications

Chemical Identifiers

1
Axypet
3 years

Specifications

±1.6/ ±0.6%
AP1000
Axypet
100 to 1000 μL
3 years
Tip Type: 1,000 μL
1
Single-Channel Pipettor
±0.4/ ±0.15%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions