Learn More
Axygen™ Axypet™ Single-Channel Pipettor
Catalog No. 14223560
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 5 mL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14223560 0.5 to 10 μL
14223569 2 to 20 μL
14223570 20 to 200 μL
14223562 100 to 1000 μL
14223573 1 to 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14223560 Supplier Axygen™ Supplier No. AP10
Only null left
Axygen™ Axypet™ pipettors combine high levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • 3-year warranty
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
Specifications
Accuracy ±4.0/ ±0.5%
Description Tip Type: 10 μL
Number of Channels 1
Product Line Axypet
Type Single-Channel Pipettor
Volume (Metric) 0.5 to 10 μL
Precision ±2.8/ ±0.4%
Warranty 3 years