Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 50μL
Clear, autoclavable tips; color-coded racks.
$1443.26 - $2592.22
Specifications
Volume (Metric) 50 μL
Color Clear
Rack Color Light Green
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
View More Specs
Products 3
Catalog Number Mfr. No. Tip Style Filtered Sterility For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Tip Style Filtered Sterility For Use With (Equipment) Price Quantity  
14-222-919
View Documents
Axygen™
ZT50R
Automation, Round Non-filtered Non-sterile Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Case of 4800 Each for $1,443.26
Only null left
14-222-920
View Documents
Axygen™
ZT50RS
Automation, Round Non-filtered Sterile Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Case of 4800 Each for $1,548.48
Only null left
14-222-926
View Documents
Axygen™
ZTF50RS
Automation, Round Filtered Sterile Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Case of 4800 Each for $2,592.22
Only null left
Description
Description
  • Clear tips
  • Light Green, Blue and Gray racks
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
50 μL
Light Green
Non-ANSI
Rack
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Polypropylene
Clear
Autoclavable
Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
Automation, Round
Caliper Zymark