missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 50μL

Catalog No. 14222919
Change view
Click to view available options
Tip Style:
Automation, Round
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
For Use With (Equipment):
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style Filtered Sterility For Use With (Equipment)
14222919 Automation, Round Non-filtered Non-sterile Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
14222920 Automation, Round Non-filtered Sterile Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
14222926 Automation, Round Filtered Sterile Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14222919 Supplier Axygen™ Supplier No. ZT50R

Clear, autoclavable tips; color-coded racks.

  • High throughput screening
  • Volume: 50μL
  • Rack Color: Light Green
  • Color: Clear
  • Designed for use with Zymark™ RapidPlate™, Zymark™ SciClone, Tecan™ Te-MO™, Tecan™ Genmate™ and Tecan™ Aquarius™ automated liquid handling systems
  • Certified RNase, DNase and Pyrogen-Free
  • Format: 96 well

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Light Green
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
Format Rack
Tip Style Automation, Round
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Product Line Caliper Zymark
Material Polypropylene
Show More Show Less