Learn More
Axygen™ Tip Refill System
Axygen™ style and universal fit pipet tips in easy-to-load refill racks
$146.96 - $178.17
Specifications
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
14-222-436
View Documents
Axygen™
RFL1200C
15 to 1250 μL
Pack of 768 Each for $166.17
Case of 6 Pack for $997.02
Only null left
14-222-437
View Documents
Axygen™
RFL222C
1 to 200 μL
Pack of 960 Each for $146.96
Case of 5 Pack for $734.80
Only null left
14-222-438
View Documents
Axygen™
RFL300C
0.1 to 10 μL
Pack of 960 Each for $178.17
Case of 5 Pack for $890.85
Only null left
Description
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
  • Specifications
    Specifications
    Non-sterile
    Clear
    Autoclavable
    Videos
    SDS
    missing translation for 'documents'
    missing translation for 'documents'