Learn More
Axygen™ Single Well Medium Profile Reagent Reservoirs
GREENER_CHOICE
Catalog No. 14222416
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14222416 Sterile
14222415 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222416 Supplier Axygen™ Supplier No. RESSW384FXSI
Get Up to $2,500 in Products with Purchase
View all promotions
Manufactured with virgin polypropylene for excellent chemical resistance
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
  • Durable and cost effective
  • Various formats for multiple applications
Specifications
Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 170 mL
Material Polypropylene
Sterility Sterile
Type Reagent Reservoir
Packaging Individually Wrapped; 25/Cs.
Channels 384
Disposable Single-use