Learn More
Axygen™ Screw Caps with O-rings
Select looped or non-looped format
Supplier:  Axygen™ SCOGR
 View more versions of this product
Catalog No. 14-222-486
Only null left
Explore available promotions
Description
Description
Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.
  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic
Tubes, with Cap, Sterile
All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.
Colored-cap versions are also available in sterile format.
Specifications
Specifications
O-ring
Gray
Non-sterile
Polypropylene
Non-looped
Both skirted and non-skirted variations of the 0.5 mL, 1.5 mL, and 2 mL microcentrifuge tubes
Product Suggestions
Product Suggestions