missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealers
Description
Axygen's PlateMax™ Semi-Automatic Plate Sealer is ideal for the low-to-medium throughput laboratory that requires uniform and consistent sealing of microplates. Offering complete versatility, the PlateMax sealer will accept a full range of plates for PCR, assay or storage applications. It can be used to seal a wide range of plate heights. Sealing parameters are set and displayed via the user-friendly control panel and the sealing operation is automated to ensure consistent results.
- Large digital display shows all operating parameters
- Adjustable sealing temperature: 100°C to 190°C
- Compatible plate materials: PP (Polypropylene), PS (Polystyrene), PE (Polyethylene)
- Compatible plate types: standard assay plates, deep well storage plates, PCR plates
Order Info
Adapter (Cat. No. 14-223-211) and heat sealing films (Cat. No. 14-223-206) sold separately.
Specifications
Specifications
| For Use With (Application) | Sealing individual plates and seals |
| No. per Pack | 1 Ea. |
| Product Type | Plate Sealer |