missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ PlateMax™ Semi Automated Plate Sealer, includes adapters for standard and deep well microplates, 230V
Description
Axygen's PlateMax™ Semi-Automatic Plate Sealer is ideal for the low-to-medium throughput laboratory that requires uniform and consistent sealing of microplates. Offering complete versatility, the PlateMax sealer will accept a full range of plates for PCR, assay or storage applications. It can be used to seal a wide range of plate heights. Sealing parameters are set and displayed via the user-friendly control panel and the sealing operation is automated to ensure consistent results.
- Large digital display shows all operating parameters
- Adjustable sealing temperature: 100°C to 190°C
- Compatible plate materials: PP (Polypropylene), PS (Polystyrene), PE (Polyethylene)
- Compatible plate types: standard assay plates, deep well storage plates, PCR plates
Specifications
Specifications
| Voltage | 230 V |
| Description | Semi-automated Plate Sealer |
| Dimensions (L x W x H) | 6.75 x 12.75 x 14.25 in. |
| No. per Pack | 1 Ea. |
| Sealing Temperature Range | 100° to 190°C |
| Weight (Metric) | 12 kg |