Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
GREENER_CHOICE
Catalog No. 14223205
Change view
Click to view available options
Product Type:
Heat Sealing Film for qPCR
Peelable Heat Sealing Film
Peelable Heat Sealing Film for qPCR
Pierceable Aluminum Heat Sealing Film
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14223205 Heat Sealing Film for qPCR
14223207 Peelable Heat Sealing Film
14223208 Pierceable Aluminum Heat Sealing Film
14223206 Peelable Heat Sealing Film for qPCR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14223205 Supplier Axygen™ Supplier No. HS100QPCR
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
Specifications
For Use With (Application) Plate Sealing
For Use With (Equipment) PlateMax Plate Sealer
Material Polyester
No. per Case 500/Cs.
No. per Pack 100/Pk.
Product Type Heat Sealing Film for qPCR