Learn More
Axygen™ PCR Tubes with 0.5 mL Flat Cap
Thin and consistent wall thickness for precise thermal transfer
$146.82 - $1,548.92
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Volume (Metric) 0.5 mL
Closure Material Polypropylene
For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
Shape Conical
View More Specs
Products 2
Catalog Number Mfr. No. Color Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Color Product Type Quantity Price Quantity & Availability  
50400540
View Documents Promotion Details
Axygen™
PCR05A
Assorted Thin Wall PCR Tube 1000/Pk., 10Pk./Cs.
Case of 11 Each for $1,548.92
Only null left
14222292
View Documents Promotion Details
Axygen™
PCR05C
Clear Thin Wall PCR Tube 1000/Pk., 10Pk./Cs.
Pack of 1000 Each for $146.82
Case of 10 Pack for $1,468.20
Only null left
Edge
Description
Description
Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.
  • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
  • Choose from a wide variety of colors
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured from polypropylene
Specifications
Specifications
RNase, DNase, pyrogen-free
Polypropylene
Conical
Polypropylene
Tube
0.5 mL
Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
Non-sterile
Flat Cap