missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PCR Tubes with 0.5 mL Flat Cap

Catalog No. 50400540
Change view
Click to view available options
Color:
Assorted
Clear
Product Type:
Thin Wall PCR Tube
Quantity:
1000/Pk., 10Pk./Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Quantity
50400540 Assorted Thin Wall PCR Tube 1000/Pk., 10Pk./Cs.
14222292 Clear Thin Wall PCR Tube 1000/Pk., 10Pk./Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50400540 Supplier Axygen™ Supplier No. PCR05A

Thin and consistent wall thickness for precise thermal transfer

Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.

  • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
  • Choose from a wide variety of colors
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured from polypropylene

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Color Assorted
Volume (Metric) 0.5 mL
Quantity 1000/Pk., 10Pk./Cs.
Closure Material Polypropylene
For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
Shape Conical
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Format Tube
Product Type Thin Wall PCR Tube
Show More Show Less