missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Multi Well Low Profile Reagent Reservoirs

Catalog No. 50400718
Change view
Click to view available options
Capacity (Metric):
32 mL
84 mL
Sterility:
Non-sterile
Sterile
Channels:
12
8
Packaging:
25/Cs.
Individually Wrapped; 25/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels Packaging
50400718 84 mL Non-sterile 12 25/Cs.
14222405 84 mL Sterile 12 Individually Wrapped; 25/Cs.
14222409 32 mL Non-sterile 8 25/Cs.
14222410 32 mL Sterile 8 Individually Wrapped; 25/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50400718 Supplier Axygen™ Supplier No. RESMW12LP

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 84 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 25/Cs.
No. per Pack 25/Pk.
Packaging 25/Cs.
Volume (Metric) Well 7 mL
Channels 12
Disposable Single-use
Show More Show Less