missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Multi Well High Profile Reagent Reservoirs

Catalog No. 50400719
Change view
Click to view available options
Capacity (Metric):
50 mL
80 mL
84 mL
264 mL
Sterility:
Non-sterile
Sterile
Channels:
12
4
8
Packaging:
25/Cs.
Individually Wrapped; 25/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels Packaging
50400719 80 mL Non-sterile 8 25/Cs.
14222403 264 mL Sterile 12 Individually Wrapped; 25/Cs.
14222406 50 mL Non-sterile 4 25/Cs.
50400716 84 mL Non-sterile 12 25/Cs.
14222408 80 mL Sterile 8 Individually Wrapped; 25/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50400719 Supplier Axygen™ Supplier No. RESMW8HP

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 80 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 25/Cs.
No. per Pack 25/Pk.
Packaging 25/Cs.
Volume (Metric) Well 10 mL
Channels 8
Disposable Single-use
Show More Show Less