Learn More
Axygen™ Eppendorf™ Compatible Pipet Tips
Compatible with Eppendorf and Biohit pipets
Supplier: Corning Incorporated TE1004BLR
Description
- Available in sterile or nonsterile forms
- e-Beam irradiated to prevent product degradation
- Maxymum recovery tips allows complete sample retrieval and greater tip-to-tip precision
- Graduation marks at 110 μL and 500 μL, allows visual confirmation of pipettor draw volume
- Volume: 1000 μL
- Color: Blue
Specifications
| 1000 μL | |
| Blue | |
| Low Retention | |
| Eppendorf | |
| 110 and 500μL |
| Non-sterile | |
| Rack | |
| Racked | |
| Yes |