missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs

Description
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.
Specifications
Specifications
| Certifications/Compliance | ANSI, RNase/DNase-free, non-pyrogenic |
| Color | White |
| Capacity (Metric) | 50 mL |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| No. per Case | 100/Cs. |
| No. per Pack | 100/Pk. |
| Packaging | Individually Wrapped; 100/Cs. |
| Disposable | Single-use |