Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs
GREENER_CHOICE
Catalog No. 14222432
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
100/Cs.
200/Cs.
Divided; 100/Cs.
Divided; 200/Cs.
Divided; Individually Wrapped; 100/Cs.
Individually Wrapped; 100/Cs.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Packaging
14222432 25 mL Sterile Individually Wrapped; 100/Cs.
14222398 25 mL Non-sterile Divided; 100/Cs.
14222399 25 mL Sterile Divided; 200/Cs.
14222400 25 mL Sterile Divided; Individually Wrapped; 100/Cs.
14222428 100 mL Sterile 200/Cs.
14222429 100 mL Sterile Individually Wrapped; 100/Cs.
14222427 100 mL Non-sterile 100/Cs.
14222430 25 mL Non-sterile 100/Cs.
14222431 25 mL Sterile 200/Cs.
14222433 50 mL Non-sterile 100/Cs.
14222434 50 mL Sterile 200/Cs.
14222435 50 mL Sterile Individually Wrapped; 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14222432 Supplier Axygen™ Supplier No. RESV25SI
Get Up to $2,500 in Products with Purchase
View all promotions
Edge
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.
Specifications
Certifications/Compliance ANSI, RNase/DNase-free, non-pyrogenic
Color White
Capacity (Metric) 25 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
Packaging Individually Wrapped; 100/Cs.
Disposable Single-use