Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs
Designed to accommodate all multichannel pipettor
Supplier:  Axygen™ RESV25
 View more versions of this product
Catalog No. 14-222-430
Only null left
Explore available promotions
Description
Description
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.
Specifications
Specifications
ANSI, RNase/DNase-free, non-pyrogenic
25 mL
Non-sterile
100/Cs.
100/Cs.
White
Polystyrene
Reagent Reservoir
100/Pk.
Single-use
Product Suggestions
Product Suggestions