Learn More
Axygen™ Biomek™ FX/NX Robotic Tips
GREENER_CHOICE
Axygen robotic tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Supplier:  Axygen™ FX38430FPRS
 View more versions of this product
Catalog No. 14222099
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.
  • Available in racked, racked sterile and with aerosol barrier (filtered)
  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications
  • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Fine Point improves accuracy and precision at lower sample volumes
  • Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents
Specifications
Chemical Identifiers
Non-filtered
Not Autoclavable
DNase, RNase and Pyrogen free
10 Racks/Pk, 5 Pks/Cs.
Polypropylene
Clear
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Beckman Biomek
Specifications
30 μL
Sterile
Not Autoclavable
DNase, RNase and Pyrogen free
Rack, 384-well
10 Racks/Pk, 5 Pks/Cs.
Beckman Biomek
Non-filtered
Clear
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Automation, Fine Point
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200
Polypropylene
Product Suggestions
Product Suggestions