missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Biomek™ FX/NX Robotic Tips

Catalog No. p-4102776
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
30 μL
45 μL
50 μL
250 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Fine Point
Automation, Round
Automation, Wide Orifice
Format:
Rack, 384-well
Rack, 96
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Tip Style Format
14222084 10 μL Non-sterile Automation, Round Rack, 96
14222090 20 μL Non-sterile Automation, Round Rack, 96
14222091 20 μL Sterile Automation, Round Rack, 96
14222098 30 μL Non-sterile Automation, Fine Point Rack, 384-well
50400366 30 μL Sterile Automation, Round Rack, 384-well
14222102 30 μL Non-sterile Automation, Round Rack, 384-well
14222099 30 μL Sterile Automation, Fine Point Rack, 384-well
14223075 45 μL Non-sterile Automation, Fine Point Rack, 384-well
14222108 50 μL Non-sterile Automation, Round Rack, 96
14222110 50 μL Non-sterile Automation, Round Rack, 96
14222111 50 μL Sterile Automation, Round Rack, 96
14222093 250 μL Sterile Automation, Round Rack, 96
14222094 250 μL Non-sterile Automation, Round Rack, 96
14222095 250 μL Sterile Automation, Round Rack, 96
14222096 250 μL Non-sterile Automation, Wide Orifice Rack, 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 14222084 Supplier Axygen™ Supplier No. FX10R
Only null left

Axygen robotic tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.

  • Available in racked, racked sterile and with aerosol barrier (filtered)
  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications
  • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Fine Point improves accuracy and precision at lower sample volumes
  • Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ANSI
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack, 96
Tip Style Automation, Round
Packaging 10 Racks/Pk, 5 Pks/Cs.
For Use With (Equipment) Beckman Coulter Biomek 3000, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200
Product Line Beckman Biomek
Material Polypropylene
Show More Show Less