missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ AxyMats™ Sealing Mats for 2mL 96 Well Assay Plates with Round Wells

Minimize evaporation in wide range of temperatures in PCR and storage applications

$169.32 - $1,058.46

Specifications

Color Natural
Material Silicone
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50/Cs.
View More Specs

Products 2
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
14222009
View Documents Promotion Details
Axygen™
AM2MLRD
Non-sterile
Pack of 10 Each for $169.32
Case of 5 Pack for $846.60
Only null left
 
14222011
View Documents Promotion Details
Axygen™
AM2MLRDS
Sterile
Case of 50 Each for $1,058.46
Only null left
 
Description

Description

Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.

Specifications

Specifications

Natural
Autoclavable
50/Cs.
Sealing Mat
96-well Microplate, 2 mL
Silicone
96
10/Pk.
Round
Videos
SDS
Documents

Documents