Learn More
Axygen™ AxyMats™ Sealing Mats for 2mL 96 Well Assay Plates with Round Wells
Catalog No. 14222009
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14222009 Non-sterile
14222011 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222009 Supplier Axygen™ Supplier No. AM2MLRD
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Color Natural
Material Silicone
Sterility Non-sterile
No. of Wells 96
No. per Case 50/Cs.
No. per Pack 10/Pk.
Product Type Sealing Mat
Plate Compatibility 96-well Microplate, 2 mL