Learn More
Axygen™ 384-well Skirted PCR Microplates
GREENER_CHOICE
Single and double notch 384-well PCR plates
Supplier:  Axygen™ PCR384M2CPBC
 View more versions of this product
Catalog No. 14224004
Only null left
Description
Description
Sealing Options: Axymats, heat sealing, adhesive films and foils.
  • Available in 50μL and 55μL
  • Single and double notch corner
Skirted (50μL)
  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Colors available by special order.
Specifications
Chemical Identifiers
RNase, DNase-free
16 x 24
384
Standard
Polypropylene
Round
Specifications
Includes Barcode
Standard
Clear
Polypropylene
384
Round
Single notch
10/Pk.
RNase, DNase-free
16 x 24
ABI real time PCR thermal cyclers and sequencers
50 μL
Non-sterile
PCR
50/Cs.
Product Suggestions
Product Suggestions