missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Wide Bore

Designed to fit a wide variety of single and multi-channel pipettes

$100.61 - $987.96

Specifications

Volume (Metric) 1 to 200 μL
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
For Use With (Equipment) Universal Fit
View More Specs

Products 2
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
14222730
View Documents Promotion Details
Axygen™
T205WBCRS
Sterile
Case of 960 Each for $987.96
Only null left
 
14222726
View Documents Promotion Details
Axygen™
T205WBC
Non-sterile
Pack of 1000 Each for $100.61
Case of 20 Pack for $2,012.20
Only null left
 
Description

Description

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

(This is the suggested replacement for T-205-WB-C-L)

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore

Order Info

Available in bulk, racked or stack rack formats.

Specifications

Specifications

1 to 200 μL
Autoclavable
Universal Fit
Clear
DNase, RNase, and Pyrogen free
Polypropylene
Videos
SDS
Documents

Documents