Learn More
Axygen™ 2.0 mL Self-Standing Screw Cap Tubes
Catalog No. 14222621
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Closure Color:
Assorted
Blue
Clear
Green
Orange
Red
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Closure Color
14222621 Sterile Assorted
14222625 Non-sterile Clear
14222624 Sterile Blue
14222626 Sterile Clear
14222627 Non-sterile Green
14222629 Sterile Green
14222631 Sterile Orange
14222630 Non-sterile Orange
14222634 Sterile Red
14222640 Sterile Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14222621 Supplier Axygen™ Supplier No. SCT200SSAS
Features chemical resistant O-rings
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Assorted
Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Material Polypropylene
Format Non-looped
Includes O-ring
Packaging Bag
Sterility Sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g
Show More Show Less