missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AXIN2 Recombinant Rabbit Monoclonal Antibody (23GB1545)
GREENER_CHOICE

Catalog No. MA552730
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552730 100 μL
MA552731 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552730 Supplier Invitrogen™ Supplier No. MA552730
Only null left

Rabbit Recombinant Monoclonal Antibody

AXIN2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

The Axin-related protein, Axin2, presumably plays an important role in the regulation of the stability of beta-catenin in the Wnt signaling pathway, like its rodent homologs, mouse conductin/rat axil. In mouse, conductin organizes a multiprotein complex of APC (adenomatous polyposis of the colon), beta-catenin, glycogen synthase kinase 3-beta, and conductin, which leads to the degradation of beta-catenin. Apparently, the deregulation of beta-catenin is an important event in the genesis of a number of malignancies. The AXIN2 gene has been mapped to 17q23-q24, a region that shows frequent loss of heterozygosity in breast cancer, neuroblastoma, and other tumors. Mutations in this gene have been associated with colorectal cancer with defective mismatch repair.

Specifications

Antigen AXIN2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1545
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AXIN2
Gene Accession No. O70240, O88566, Q9Y2T1
Gene Alias Axi1; axil; axin 2; Axin like protein; axin2; axin-2; Axin-like protein; Axis inhibition protein 2; conductin; DKFZp781B0869; MGC10366; MGC126582; ODCRCS
Gene Symbols AXIN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Axin2 (58-93AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12006, 29134, 8313
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less