Learn More
Invitrogen™ Avidin Protein, HRP
Supplier:  Invitrogen™ 29994
 View more versions of this product
Catalog No. PI29994
Only null left
Explore available promotions
Description
Description
Avidin from chicken egg whites is a glycoprotein that effectively binds biotin, a tag that is frequently used to tag antibodies and other probes for immunodetection methods. Pure avidin is a key component in certain immunohistochemistry (IHC) methods. Alkaline phosphatase (AP) and horseradish peroxidase (HRP) conjugates of avidin are useful for Western blotting or ELISA detection with appropriate enzyme substrates.
Highlights:
  • Native avidin – glycoprotein from chicken egg whites, 67kDa, pI equals 10
  • Soluble – glycosylation and isoelectric point renders avidin more soluble than streptavidin
  • Affordable – significantly less expensive than recombinant streptavidin
  • ABC staining – avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
  • Pure protein and conjugates – choose pure, unconjugated avidin for use in ABC staining or peroxidase or alkaline phosphatase forms for membrane and plate-based assays
Specifications
Specifications
5 mg
Store at -20°C. Shipped at ambient temperature.
HRP (Horseradish Peroxidase)
Western Blotting, ELISA, Immunohistochemistry
HRP (Horseradish Peroxidase)
Biotin
Biotin
Solid
Pierce™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.