missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Avidin, NeutrAvidin™, Oregon Green™ 488 conjugate

Catalog No. A6374
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A6374 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A6374 Supplier Invitrogen™ Supplier No. A6374

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Avidin, NeutrAvidin™, Oregon Green™ 488 conjugate

Our Oregon Green™ 488 conjugate of NeutrAvidin™ biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, green-fluorescent reagent (absorption/emission maxima ∼496/524 nm) is ideal for detecting biotinylated targets in fluorescence microscopy and flow cytometry applications.

Specifications

Conjugate Oregon Green 488
Quantity 1 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Line NeutrAvidin, Oregon Green

For Research Use Only. Not for use in diagnostic procedures.