missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Avidin, HRP conjugate
Description
Reactivity: HRP-Avidin reacts with biotin.
Applications: Immunohistochemistry, ELISA
Specifications
Specifications
| Conjugate | HRP |
| Quantity | 5 mg |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Wet Ice |
| Form | Liquid |
| Label or Dye | HRP (Horseradish Peroxidase) |
Frequently Asked Questions (FAQs)
Can you provide the shelf life for Avidin, HRP conjugate (Cat. No. 434423)?
The Avidin, HRP conjugate (Cat. No. 434423) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).
For Research Use Only. Not for use in diagnostic procedures.