missing translation for 'onlineSavingsMsg'
Learn More

Avenanthramide C, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012658
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012658 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012658 Supplier Merck Emd Millipore Supplier No. 3646510MG

Avenanthramides are a class of substituted N-cinnamoylanthranilic acids present mainly in oats.

Specifications

CAS 116764-15-9
Physical Form Neat
Linear Formula C16H13 NO6
MDL Number MFCD23380831
Synonym 2-{[(2 E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]amino}-5-hydroxybenzoic acid; N-[3 ′,4 ′-Dihydroxy-(E)-cinnamoyl]-5-hydroxyanthranilic acid; Avenanthramide Bc
Type Analytical Standard
Quantity 10 mg
Formula Weight 315.28
Percent Purity ≥98.0% (GC)
Grade Analytical Standard