missing translation for 'onlineSavingsMsg'
Learn More

Avenanthramide B, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002713
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002713 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002713 Supplier Merck Emd Millipore Supplier No. 9310510MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 108605-69-2
Physical Form Neat
Linear Formula C17H15 NO6
MDL Number MFCD23380832
Synonym 5-Hydroxy-2-{[(2 E)-3-(4-hydroxy-3-methoxyphenyl)-1-oxo-2-propenyl]amino}benzoic acid; N-[4′-Hydroxy-3′-methoxy-(E)-cinnamoyl]-5-hydroxyanthranilic acid; Avenanthramide Bf
Type Analytical Standard
Quantity 10 mg
Formula Weight 329.30
Percent Purity ≥98% (HPLC)
Grade Analytical Standard