missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Autosampler Installation Kit (Septa), 384-well plate
Description
The 384-Well Plate Autosampler Kit (for use with septa) for 3730 and 3730xl DNA Analyzer. This kit includes the plate base, retainer, septa and 384-well microplates. This system enables plates to be attached to the instrument autosampler.
Specifications
Specifications
| Content And Storage | This kit includes the plate base, retainer, septa and 384-well microplates. |
| Format | 384-well Plate |
| For Use With (Equipment) | 3730 DNA Analyzer |
| For Use With (Application) | Sequencing |
| Product Type | Autosampler Installation Kit |
| Quantity | 1 Kit |
For Research Use Only. Not for use in diagnostic procedures.