missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Autophagy Inhibitor, 3-MA

CAS: 5142-23-4 | C6H7N5, C6H7N5 | 149.16 g/mol

Supplier:  Thermo Scientific Chemicals J64813MF

 View more versions of this product

Catalog No. AAJ64813MF


Only null left

Description

Description

A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
Specifications

Specifications

50 mg
2°C to 8°C
5142-23-4
149.15g/mol
Signal Transduction Reagent
MFCD00010531
FSASIHFSFGAIJM-UHFFFAOYSA-N
3-methyl-3H-purin-6-amine
White
A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
C6H7N5
3-Methyladenine; 6-Amino-3-methylpurine
Solid
146087
CN1C=NC(N)=C2N=CN=C12
149.16

Chemical Identifiers

5142-23-4
C6H7N5
3-Methyladenine; 6-Amino-3-methylpurine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 225-908-6

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only