Learn More
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.
$525.54 - $1480.46
Specifications
| Color | Clear |
|---|---|
| Format | Rack |
| For Use With (Equipment) | Eppendorf epMotion |
| Product Line | Eppendorf epMotion |
| Catalog Number | Mfr. No. | Volume (Metric) | Filtered | Sterility | Fit for Purpose | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Filtered | Sterility | Fit for Purpose | Price | Quantity | ||
|
14-754-201
|
N/A | 50 μL | Filtered | Sterile | Sterile and PCR Ready |
Pack of 960 Each for $1,480.46
Case of 4 Pack for $5,921.84
|
|
||
|
14-754-202
|
N/A | 50 μL | Non-filtered | Sterile | Sterile |
Pack of 960 Each for $575.39
Case of 4 Pack for $2,301.56
|
|
||
|
14-754-203
|
N/A | 50 μL | Non-filtered | Non-sterile | Certified Quality |
Pack of 960 Each for $525.54
Case of 4 Pack for $2,102.16
|
|
||
|
14-754-204
|
N/A | 300 μL | Filtered | Sterile | Sterile and PCR Ready |
Pack of 960 Each for $1,103.18
Case of 4 Pack for $4,412.72
|
|
||
|
14-754-205
|
N/A | 300 μL | Non-filtered | Sterile | Sterile |
Pack of 960 Each for $575.39
Case of 4 Pack for $2,301.56
|
|
||
|
14-754-206
|
N/A | 300 μL | Non-filtered | Non-sterile | Certified Quality |
Pack of 960 Each for $666.25
Case of 4 Pack for $2,665.00
|
|
||
|
14-754-207
|
N/A | 1000 μL | Filtered | Sterile | Sterile and PCR Ready |
Pack of 960 Each for $1,138.89
Case of 4 Pack for $4,555.56
|
|
||
|
14-754-208
|
N/A | 1000 μL | Non-filtered | Sterile | Sterile |
Pack of 960 Each for $575.39
Case of 4 Pack for $2,301.56
|
|
||
|
14-754-209
|
N/A | 1000 μL | Non-filtered | Non-sterile | Certified Quality |
Pack of 960 Each for $533.89
Case of 4 Pack for $2,135.56
|
|
||
Description
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:- Available as sterile and filtered.
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
- Lot-tested during production on actual workstation for which they are created.
- Low percentage coefficient of variation (%CV).
Warranty:A five (5)-year shelf life from the date of manufacture is recommended.
Specifications
| Clear | |
| Rack | |
| Eppendorf epMotion | |
| Eppendorf epMotion |