missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
GREENER_CHOICE

Catalog No. 14754201
Change view
Click to view available options
Volume (Metric):
50 μL
300 μL
1000 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility Fit for Purpose
14754201 50 μL Filtered Sterile Sterile and PCR Ready
14754202 50 μL Non-filtered Sterile Sterile
14754203 50 μL Non-filtered Non-sterile Certified Quality
14754204 300 μL Filtered Sterile Sterile and PCR Ready
14754205 300 μL Non-filtered Sterile Sterile
14754206 300 μL Non-filtered Non-sterile Certified Quality
14754207 1000 μL Filtered Sterile Sterile and PCR Ready
14754208 1000 μL Non-filtered Sterile Sterile
14754209 1000 μL Non-filtered Non-sterile Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14754201

Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.

Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.

Features:

  • Available in nonsterile, sterile and sterile filtered
  • Validated on actual workstation for which tip is created
  • Low percentage coefficient of variation
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen

Specifications

Volume (Metric) 50 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Eppendorf epMotion
Product Line Eppendorf epMotion
Fit for Purpose Sterile and PCR Ready