Learn More
Thermo Scientific™ Automation Tips for Qiagen™ Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Supplier: Thermo Scientific™ 951021
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
- Available in nonsterile, sterile and with the ART™ filter technology
- Lot-tested during production on actual workstation for which they are created
- Low percentage coefficient of variation (%CV)
Specifications
| 250 μL | |
| Sterile | |
| Rack | |
| 96 Tips/Tray, 10 Trays/Case | |
| Qiagen, Qiagen |
| Filtered | |
| Clear | |
| Automation | |
| Universal Fit, Qiagen | |
| Sterile and PCR Ready |