Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.
Supplier:  Thermo Scientific™ 91826205
 View more versions of this product
Catalog No. 21-402-637
Only null left
Explore available promotions
Description
Description
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
Specifications
Specifications
20 μL
Non-sterile
Autoclavable
Free of RNase, DNase, DNA, ATP, and endotoxin
Low Retention
4800 tips
Beckman
Non-filtered
Clear
ISO 9001 and ISO 13485
Rack
96 tips/rack, 10 racks/pack, 5 packs/case
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96
Certified Quality
Product Suggestions
Product Suggestions