missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Automated Thermal Cycler (ATC) + Plastics Packages, 96-well

Catalog No. A41199
Encompass
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Description:
Includes 2 x MicroAmp EnduraPlate Optical 96-well Full-Skirted Clear Reaction Plates with Barcode (100 plates total)
Includes 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
A41199 Includes 2 x MicroAmp EnduraPlate Optical 96-well Full-Skirted Clear Reaction Plates with Barcode (100 plates total)
A41198 Includes 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. A41199 missing translation for 'mfr' Applied Biosystems™ missing translation for 'supplierNo' A41199

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For customers seeking to automate their PCR workflow, the Automated Thermal Cycler (ATC) provides the reliability and performance you’ve come to expect from Applied Biosystems instruments.

For customers seeking to automate their PCR workflow, the Automated Thermal Cycler (ATC) provides the reliability and performance you’ve come to expect from Applied Biosystems instruments. Now in a small, easy-to-integrate, and compatible format, our Automated Thermal Cycler enables hands-free PCR results.

This Automated Thermal Cycler (ATC) package includes a thermal cycler plus MicroAmp EnduraPlates (full-skirt or half-skirt) and MicroAmp Clear Adhesive Films.

Automated Thermal Cycler
• Space-saving design with our smallest thermal cycler footprint
• Flexible modular design helps save space on a robotic deck
• Separate control panel module for flexibility when integrating on a robotic deck
• Choice of 10-cm, 1-m, and 3-m cables for flexibility to locate control module under thermal cycler, under deck, or up to 3 meters away
• Automated lid compatible with programming for easy, hands-free operation with a liquid handler or plate stacker
• Heated cover slides forward to cover plate nest to help ensure thermal contact and eliminate evaporation
• Compatible with full-skirt or half-skirt plates

MicroAmp EnduraPlates
MicroAmp EnduraPlates offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. They are a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments. They are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.
• Optimized for performance with Applied Biosystems instruments
• Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete workflow needs

MicroAmp Clear Adhesive Films
MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.
• Adhesive provides tight seal over wells to prevent leaks and evaporation.
• Easy to apply and remove
• Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky)

Specifications

Thermal Accuracy ±0.25°C (35°C to 99.9°C)
Content And Storage • Automated Thermal Cycler (ATC), 96-well, 1-meter cable
• 2 x MicroAmp EnduraPlate Optical 96-well Full-Skirted Clear Reaction Plates with Barcode (100 plates total)
• 1 x MicroAmp Clear Adhesive Films (100 films)

High-throughput Compatibility High-throughput Compatible
Thermal Uniformity <0.5°C (20 sec. after reaching 95°C)
Reaction Speed Standard
Dimensions 12.5 x 7.1 x 5.3 in. (31.7 x 17.9 x 13.3 cm)
Capacity 96-well, 0.2 mL, full-skirted plate
Voltage 100/240 V
Display Type No Display
Product Type Thermal Cycler
Warranty Limited Warranty
Thermal Range 4°C to 105°C
User Interface USB, Wi-Fi, Ethernet
Wattage 600 W
Max. Ramp Rate 3.5°C/sec. (Block), 1.8°C/sec. (Sample)
Description Includes 2 x MicroAmp EnduraPlate Optical 96-well Full-Skirted Clear Reaction Plates with Barcode (100 plates total)
Quantity 1 package
Weight 13.2 lb. (6.0 kg)
Block Configurations 96-well Isothermal Block
Show More Show Less
Do any of your thermal cyclers, other than the Automated Thermal Cycler, hold a full skirted PCR plate?

For full skirt plate, a raised heating block is required. None of our thermal cyclers have a raised heating block except the Automated Thermal Cycler.

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf