Learn More
Applied Biosystems™ AutoMate Express™ Nucleic Acid Extraction System
Catalog No. 361055934
Change view
Click to view available options
Quantity:
1 instrument
Catalog No. Quantity
361055934 1 instrument
1 options
Catalog No. 361055934 Supplier Applied Biosystems™ Supplier No. 4467754
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The AutoMate Express Nucleic Acid Extraction System is an easy-to-use benchtop instrument that enables hands-free automation of the nucleic acid extraction process for contaminant and impurity testing.
The AutoMate Express Nucleic Acid Extraction System is an easy-to-use benchtop instrument that enables hands-free automation of the nucleic acid extraction process for contaminant and impurity testing. The instrument uses the established PrepSEQ nucleic acid extraction chemistry for quantitative recovery of DNA and RNA from diverse samples types, including cell culture, raw material (i.e., serum), and in-process downstream purification samples such as column purification or bulk drug substance (BDS) samples.
Features of the AutoMate Express Nucleic Acid Extraction System include:
  • Time-saving, cost-effective DNA extraction for mycoplasma, MMV, and vesivirus
  • Reproducible recovery of DNA from complex samples
  • Out-of-the-box automation with prefilled PrepSEQ Express cartridges and ready-to-run protocols
  • Closed system minimizes the risk of contamination
  • Ideal for in-process testing of contaminants
Three-in-one extraction of cell culture contaminants
The AutoMate Express system can be used to perform nucleic acid extraction of the common contaminants in mammalian cell culture manufacturing, i.e., mycoplasmas, MMV, and vesiviruses (Figure 1). Flexible protocols are available to isolate DNA and RNA from a single organism, or a 3-in-1 surveillance panel protocol can simultaneously extract mycoplasma, MMV, and vesivirus nucleic acid from a single sample.
Closed system with walk-away automation
The AutoMate Express system automatically performs all steps for efficient nucleic acid extraction from contaminants in a closed environment. The system uses PrepSEQ Express kits that contain sealed cartridges with extraction reagents, and plastics, including tips and sample, lysis, and elution tubes. The workflow has been designed for a completely hands-free operation enabling reproducible extraction efficiencies and reduced time-to-results.
Specifications
Product Type Nucleic Acid Extraction System
For Use With (Application) Quantitative recovery of DNA and RNA
Product Line AutoMate Express™
Quantity 1 instrument
Samples per Run Up to 13 samples
For Research Use Only. Not for use in diagnostic procedures.