Learn More
Keystone Cleanroom Products Autoclavable Tyvek™ Breather Bags with Self-Sealing Closure
These Tyvek™ Breather Bags will not rip or puncture. Includes self-sealing closure.
$1161.57 - $7391.83
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | Certificate of Conformance (COC) |
| Closure Type | Self-seal |
| Color | Clear, White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity | ||||
|
01-100-363
|
Keystone Cleanroom Products
00206TWBBSSSD |
6 in. | 2 in. |
Case of 1000 Each for $2,232.63
|
|
||||
|
01-100-342
|
Keystone Cleanroom Products
00209TWBBSSSD |
9 in. | 2 in. |
Case of 1000 Each for $2,549.42
|
|
||||
|
01-100-400
|
Keystone Cleanroom Products
00306TWBBSSSD |
6 in. | 3 in. |
Case of 1000 Each for $2,323.14
|
|
||||
|
01-100-387
|
Keystone Cleanroom Products
00308TWBBSSSD |
8 in. | 3.5 in. |
Case of 500 Each for $1,161.57
|
|
||||
|
01-100-391
|
Keystone Cleanroom Products
00415TWBBSSSD |
15 in. | 4 in. |
Case of 500 Each for $2,458.91
|
|
||||
|
01-100-331
|
Keystone Cleanroom Products
00422TWBBSSSD |
22 in. | 4 in. |
Case of 500 Each for $2,458.91
|
|
||||
|
01-100-392
|
Keystone Cleanroom Products
00510TWBBSSSD |
10 in. | 5 in. |
Case of 500 Each for $1,795.15
|
|
||||
|
01-100-351
|
Keystone Cleanroom Products
00608TWBBSSSD |
8 in. | 6 in. |
Case of 500 Each for $1,719.73
|
|
||||
|
01-100-395
|
Keystone Cleanroom Products
00612TWBBSSSD |
12 in. | 6 in. |
Case of 500 Each for $2,458.91
|
|
||||
|
01-100-396
|
Keystone Cleanroom Products
00628TWBBSSSD |
28 in. | 6 in. |
Case of 500 Each for $3,982.53
|
|
||||
|
01-100-316
|
Keystone Cleanroom Products
00632TWBBSSSD |
32 in. | 6 in. |
Case of 500 Each for $6,019.05
|
|
||||
|
01-100-353
|
Keystone Cleanroom Products
00635TWBBSSSD |
35 in. | 6 in. |
Case of 500 Each for $6,335.86
|
|
||||
|
01-100-354
|
Keystone Cleanroom Products
00713TWBBSSSD |
13 in. | 7 in. |
Case of 500 Each for $2,564.51
|
|
||||
|
01-100-364
|
Keystone Cleanroom Products
00810TWBBSSSD |
10 in. | 8 in. |
Case of 500 Each for $2,564.51
|
|
||||
|
01-100-381
|
Keystone Cleanroom Products
00815TWBBSSSD |
15 in. | 8 in. |
Case of 500 Each for $2,971.81
|
|
||||
|
01-100-329
|
Keystone Cleanroom Products
00818TWBBSSSD |
18 in. | 8 in. |
Case of 500 Each for $3,846.76
|
|
||||
|
01-100-344
|
Keystone Cleanroom Products
00834TWBBSSSD |
34 in. | 8 in. |
Case of 500 Each for $5,626.84
|
|
||||
|
01-100-328
|
Keystone Cleanroom Products
01011TWBBSSSD |
11 in. | 10 in. |
Case of 500 Each for $3,545.06
|
|
||||
|
01-100-373
|
Keystone Cleanroom Products
01020TWBBSSSD |
20 in. | 10 in. |
Case of 500 Each for $4,208.80
|
|
||||
|
01-100-388
|
Keystone Cleanroom Products
01214TWBBSSSD |
14 in. | 12 in. |
Case of 500 Each for $3,982.53
|
|
||||
|
01-100-397
|
Keystone Cleanroom Products
01217TWBBSSSD |
17 in. | 12 in. |
Case of 500 Each for $4,314.41
|
|
||||
|
01-100-399
|
Keystone Cleanroom Products
01424TWBBSSSD |
24 in. | 14 in. |
Case of 500 Each for $7,195.71
|
|
||||
|
01-100-398
|
Keystone Cleanroom Products
01520TWBBSSSD |
20 in. | 15 in. |
Case of 500 Each for $6,003.98
|
|
||||
|
01-100-393
|
Keystone Cleanroom Products
01617TWBBSSSD |
17 in. | 16 in. |
Case of 500 Each for $5,913.46
|
|
||||
|
01-100-335
|
Keystone Cleanroom Products
01626TWBBSSSD |
26 in. | 16 in. |
Case of 500 Each for $7,391.83
|
|
||||
|
01-100-320
|
Keystone Cleanroom Products
01632TWBBSSSD |
32 in. | 16 in. |
Case of 250 Each for $4,812.22
|
|
||||
|
01-100-336
|
Keystone Cleanroom Products
01635TWBBSSSD |
35 in. | 16 in. |
Case of 250 Each for $5,460.89
|
|
||||
|
01-100-394
|
Keystone Cleanroom Products
01822TWBBSSSD |
22 in. | 18 in. |
Case of 500 Each for $7,316.39
|
|
||||
|
01-100-310
|
Keystone Cleanroom Products
02426TWBBSSSD |
26 in. | 24 in. |
Case of 250 Each for $5,038.50
|
|
||||
|
01-100-389
|
Keystone Cleanroom Products
02430TWBBSSSD |
30 in. | 24 in. |
Case of 250 Each for $5,294.96
|
|
||||
|
01-100-324
|
Keystone Cleanroom Products
02435TWBBSSSD |
35 in. | 24 in. |
Case of 250 Each for $5,657.00
|
|
||||
|
01-100-384
|
Keystone Cleanroom Products
X0308TWBBSSSD |
8 in. | 3 in. |
Case of 500 Each for $1,161.57
|
|
||||
Description
The medical grade 1073B Tyvek™ material has the highest microbial barrier properties of any steam sterilization compatible materials. Tyvek is also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
- Manufactured and packaged in an ISO certified cleanroom
- Chevron Peel Opening
- Autoclavable at up to 125°C
- Tear and puncture resistant
- Hydrophobic
- Lint-free
- Convenient Self-Sealing Closure
Specifications
| Autoclavable | |
| Self-seal | |
| Single-use | |
| Tyvek | |
| Yes |
| Certificate of Conformance (COC) | |
| Clear, White | |
| Steam, EtO, Gamma or VHP Sterilization | |
| Non-sterile | |
| Autoclave Bag |